Anti-ADAM17 Chicken Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF930
220014-473EA 698.49 USD
220014-473
Anti-ADAM17 Chicken Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ADAM17
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR