Anti-IL12 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF912
220014-466EA 753.45 USD
220014-466
Anti-IL12 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: IL12
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR