Anti-CSF2RB Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF906
220014-460EA 698.49 USD
220014-460
Anti-CSF2RB Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CSF2RB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR