Anti-GAS6 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF885
220014-455EA 746.22 USD
220014-455
Anti-GAS6 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GAS6
Antigen Name: growth arrest-specific 6
Antigen Synonyms: AXLLG, AXSF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR