Anti-CCL19 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF880
220014-453EA 746.22 USD
220014-453
Anti-CCL19 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CCL19
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR