Anti-IGFBP1 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF871
220014-447EA 746.22 USD
220014-447
Anti-IGFBP1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: IGFBP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR