Anti-CXCL14 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF866
220014-445EA 698.49 USD
220014-445
Anti-CXCL14 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CXCL14
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR