Anti-CDH3 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF861
220014-444EA 714.4 USD
220014-444
Anti-CDH3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CDH3
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR