Anti-GDF1 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF858
220014-441EA 698.49 USD
220014-441
Anti-GDF1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GDF1
Antigen Name: growth differentiation factor 1
Antigen Synonyms: AI385651, Gdf-1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR