Anti-GDF5 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF853
220014-437EA 698.49 USD
220014-437
Anti-GDF5 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GDF5
Antigen Name: growth differentiation factor 5
Antigen Synonyms: BMP-14, Cdmp-1, bp, brp
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR