Anti-EEA1 Sheep Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF8047
102879-106EA 714.4 USD
102879-106
Anti-EEA1 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: EEA1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR