Anti-CD70 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF783
220014-403EA 698.49 USD
220014-403
Anti-CD70 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CD70
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR