You Searched For: Staples
Staples are used for binding materials together in both office and heavier industrial uses. Large staples might be used with a hammer or staple gun for masonry, roofing, corrugated boxes, and other heavy-duty uses. These tools boast superior puncturing power for easy application with soft and hard surfaces, ensuring tight and securing binding. Most models are universally compatible with everyday office staplers, allowing for easy pairing without special models being required.
Staples are used for binding materials together in both office and heavier industrial uses. Large staples might be used with a hammer or staple gun for masonry, roofing, corrugated boxes, and other heavy-duty uses. These tools boast superior puncturing power for easy application with soft and hard surfaces, ensuring tight and securing binding. Most models are universally compatible with everyday office staplers, allowing for easy pairing without special models being required.
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Janitorial Supplies
Description: These heavy-duty staples are ideal for stapling jobs such as insulation, carpet underlaying and roofing felt.
Supplier: Janitorial Supplies
Description: Premium-quality carbon steel staples are designed specifically for Bostitch® heavy-duty staplers
Catalog Number: (500025-693)
Supplier: Janitorial Supplies
Description: The ultimate in Swingline® staple performance for Optima® staplers
Supplier: Janitorial Supplies
Description: Specially engineered to staple up to 50% more paper volume
Catalog Number: (500025-653)
Supplier: Janitorial Supplies
Description: Economical staples help keep you within budget
Supplier: Janitorial Supplies
Description: Color-coded staple cartridge makes it easy to identify and keep supplies organized.
Catalog Number: (500025-676)
Supplier: Janitorial Supplies
Description: 100% Performance Guarantee! The ultimate Swingline® staple performance for standard staplers
Catalog Number: (500025-673)
Supplier: Janitorial Supplies
Description: High capacity staples feature stronger, thinner wire that produces a streamlined appearance ideal for presentation pieces
Supplier: Janitorial Supplies
Description: Rely on top-quality, heavy-duty staples for clean, consistent stapling again and again
Catalog Number: (500042-663)
Supplier: Janitorial Supplies
Description: High-capacity flat-clinch staples are designed for use with Rapid S50.
Catalog Number: (500025-695)
Supplier: Janitorial Supplies
Description: Rely on these high-quality standard staples to give you consistent, dependable, easy stapling
Supplier: Janitorial Supplies
Description: Save time and money by stapling up to 5,000 times without reloading
Supplier: Janitorial Supplies
Description: Enjoy increased efficiency with this easy-loading cartridge that staples up to 5,000 times without reloading.
Catalog Number: (500025-639)
Supplier: Janitorial Supplies
Description: 100% Performance Guarantee! The ultimate Swingline® staple performance for standard staplers
Catalog Number: (500025-694)
Supplier: Janitorial Supplies
Description: With chisel point tips for maximum penetration power.
Catalog Number: (500025-643)
Supplier: Janitorial Supplies
Description: Streamline your workday with this staple cartridge that adds speed and convenience to your fastening tasks.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 19
no targeter for Bottom