Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless Steel Acid Cabinet Base Units, Kloppenberg & Co.

Supplier: KLOPPENBERG & CO.
BAC242235 BAC482235 BAC362235
These cabinets are comprised of quality T304 stainless steel

The single-door unit has a #4 brushed satin finish. There are one or two hinged, louvered doors and they are designed with an "acid" cabinet labeling and spill containment trays. As with all Kloppenberg products, the stainless steel used is made of an industry standard minimum of 80% recyclable materials, ensuring this product is not only long lasting but eco-friendly.

Certifications: SEFA 11 Compliant and SEFA Section 8 tested and approved.

Ordering information: Can be adjusted to meet ADA height requirements. Type 316 available for additional pricing.



 

ORDER

Expand All / Collapse All
Description Depth Height Width Supplier No. VWR Catalog Number Unit Price Quantity
Double Door Base Unit 22" 343/4" 36" BAC362235 10039-582 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Double Door Base Unit 22" 35" 48" BAC482235 10039-584 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Single Door Base Unit 22" 343/4" 24" BAC242235 10039-580 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty