Stainless Steel Solid Hinged Door Wall Unit, Kloppenberg & Co.
WB241330 WB181330
10039-586EA 1043.15 USD
10039-586 10039-568
Stainless Steel Solid Hinged Door Wall Unit, Kloppenberg & Co.
Stainless Steel Solid Hinged Door Wall Unit crafted in quality type 304 stainless steel
There is a single adjustable shelf and a solid back. As with all Kloppenberg products, the stainless steel used is made of an industry standard minimum of 80% recyclable materials, ensuring this product is not only long lasting but eco-friendly.
Certifications: SEFA Section 8 tested and approved.