Combination Lab Refrigerator/ Freezer, General Purpose, Thermo Scientific
Supplier: Thermo Scientific
MC20SS-SAEE-TS MC20SS-SAEE-TS
89500-302EA 20320.44 USD
CA89500-302 89500-302
Combination Lab Refrigerator/ Freezer, General Purpose, Thermo Scientific
Freezers Laboratory / General Purpose Freezers
General Purpose (GP) Series Combination Lab Refrigerator/Freezer for routine laboratory applications.
  • Dual refrigeration system
  • Digital temperature controller for each chamber with intuitive interface
  • Temperature stability of ±3°C
  • High-density, fluorine-free insulation with automatic defrost
  • With keyed on/off switch and High/low temperature; audio/visual alarms
  • Easily accessible contacts for backup monitoring with door ajar alarm and lockable controller settings
High-quality steel exterior front and aluminum interior and casters for easy mobility and Quiet refrigeration system.
Certifications: cUL listed.