Single Element ICP and ICP/MS Certified Reference Standards, Rhenium, ARISTAR®, VWR Chemicals BDH®
BDHCGRE1-1 BDHCGRE1-1
82026-042EA 93.83 USD
CA82026-042 82026-042
Single Element ICP and ICP/MS Certified Reference Standards, Rhenium, ARISTAR®, VWR Chemicals BDH®
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online. Most products are available in stock at the 125mL and 500mL sizes packaged in plastic LDPE bottles.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 ppm.