Jackson Original Tracheostomy Tube, Disposable Grade, Sklar
96-2806 96-2809 96-2801 96-2802 96-2800 96-2807 96-2812 96-2805 96-2804 96-2808 96-2810 96-2811 96-2803
10185-880CS 3519.92 USD
10185-880 10185-890 10185-884 10160-090 10185-874 10185-882 10185-872 10160-094 10185-888 10185-878 10185-886 10160-092 10185-876
Jackson Original Tracheostomy Tube, Disposable Grade, Sklar
Sklar's Jackson Original Style Tracheostomy Tube consists of one obturator, an inner cannula, and an outer cannula.
These three pieces work together to ensure that you can breathe properly following a tracheal incision. The obturator is used to safely and gently place the outer cannula into the incision and into the trachea. The outer cannula is the part of the device which keeps the actual incision in the neck open. The neck plate, which is attached to the outer cannula, is where the side slots are located for tying or strapping the tube around the patient's neck, as well as part of the locking mechanism. Once the outer cannula is secured, the obturator is removed and the inner cannula is inserted. The Jackson original style tube features a lock on the faceplate that protrudes upward slightly and swivels over the inner cannula to secure it in place. The original style inner cannula is also designed with "wings" on each side that are used for its removal.