Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Medline Professional® Nitrile Exam Gloves

Supplier: MEDLINE INDUSTRIES
PRO31761 PRO31760 PRO31763 PRO31762 PRO31764
Medline Professional™ Nitrile Exam Gloves with Aloe are the standard in protection and comfort.

  • Designed specifically with the lab environment in mind
  • Tested for use with chemotherapy drugs
  • Thickest Professional Nitrile gloves
  • All-over texturing for better grip on small or large items, wet or dry

Gloves are used for protection against Chemotherapy Drugs, so exposure must be selected specifically for the type of Chemotherapy agent used.



 

ORDER

Expand All / Collapse All
Description Glove Size Length Thickness Glove Color Supplier No. VWR Catalog Number Unit Price Quantity
Medline Professional Nitrile X-Small 24.1 cm (9 1/2") 0.15 mm Green PRO31760 10053-706 Case of 1000 Retrieving
Pack of 100 Retrieving
Medline Professional Nitrile Small 24.1 cm (9 1/2") 0.15 mm Green PRO31761 10053-708 Case of 1000 Retrieving
Pack of 100 Retrieving
Medline Professional Nitrile Medium 24.1 cm (9 1/2") 0.15 mm Green PRO31762 10053-710 Case of 1000 Retrieving
Pack of 100 Retrieving
Medline Professional Nitrile Large 24.1 cm (9 1/2") 0.15 mm Green PRO31763 10053-712 Case of 1000 Retrieving
Pack of 100 Retrieving
Medline Professional Nitrile X-Large 24.1 cm (9 1/2") 0.15 mm Green PRO31764 10053-714 Case of 1000 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/Hand-specific Ambidextrous
Disposable/Reusable Disposable
Powdered/Powder-free Powdered
Cuff Style Beaded
Finger Thickness 5.9 mil
Palm Thickness 5.5 mil

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty