Pyridine ACS, VWR Chemicals BDH®

Danger

BDHVBDH4562-1L
BDH4562-1LEA 297.79 USD
BDH4562-1L
Pyridine ACS, VWR Chemicals BDH®
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 12,08153
https://us.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg

Order Now

Specification Test Results

Assay Min. 99.0 %
Reducing substances Passes test
Solubility in water Passes test
Ammonia Max. 0.002 %
Residue after evaporation Max. 0.002 %
Water Max. 0.1 %
Cl (Chloride) Max. 0.001 %
SO4 (Sulphate) Max. 0.001 %
Cu (Copper) Max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR