Copper(II) nitrate hemipentahydrate, crystallised ACS, VWR Chemicals BDH®

Danger

Synonyms: Cupric Nitrate HydrateCupric nitrate hemipentahydrateCopper dinitrate hemipentahydrateCupric nitrate 2.5-hydrateCupric nitrate hemi(pentahydrate)

BDHVBDH4558-500G
BDH4558-500GCS 979.52 USD
BDH4558-500G
Copper(II) nitrate hemipentahydrate, crystallised ACS, VWR Chemicals BDH®
Copper (II) nitrate hemipentahydrate
Formula: Cu(NO₃)₂·2.5H₂O
MW: 232.59 g/mol
Melting Pt: 255 °C
Density: >1 g/cm³ (20 °C)
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Insoluble matter Max. 0.01 %
Cl (Chloride) Max. 0.002 %
SO4 (Sulphate) Max. 0.01 %
Ca (Calcium) Max. 0.005 %
Fe (Iron) Max. 0.005 %
K (Potassium) Max. 0.005 %
Na (Sodium) Max. 0.01 %
Ni (Nickel) Max. 0.01 %
Pb (Lead) Max. 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR