Would you like to log in again?
5' Purifier® Logic®+ Class II A2 Biosafety Cabinets, International Electrical Configurations, Labconco®
Supplier: Labconco
302511150 302511030 302511130 302510140 302511070 302510040 302511170 302511050 302580130 302581120 302581020 302580150 302581140 302580030 302581040 302580170 302580050 302580070 302511040 302510150 302511140 302510070 302510130 302510030 302510050 302510170 302580020 302580120 302580040 302581130 302581030 302580140 302581150 302581050 302581170 302581070
The new Purifier Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality.
  • Intuitive programming
  • Color-coded menus
  • Status messaging and alarms
  • Diagnostic troubleshooting
  • Provides personnel, product and environmental protection
  • Suitable for work in biosafety levels 1, 2 and 3
Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system
Using the same ECM blower technology as the Purifier Logic®, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. These biohazard cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. In addition, some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. Please refer to sash opening, electrical configuration and product specifications for details
MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.
Accessories information: Replacement supply HEPA filter for Purifier Logic Biosafety Cabinets and Purifier Cell Logic Biosafety Cabinets. HEPA Filters are 99.99% efficient (with particles 0.3 micron) and have aluminum frames with either aluminum separators or separatorless pleats.
ORDER
Expand All / Collapse All
Description Electrical Sash Opening Supplier No. VWR Catalog Number Unit Price Quantity
5' Purifier Logic+ A2 Biosafety Cabinet 100V Japan 8" 302580020 89412-634 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V UK 8" 302580050 89412-682 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V UK 10" 302510050 89412-690 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V Schuko 8" 302580030 89412-746 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V Schuko 10" 302510030 89412-754 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V China/Aust 8" 302580040 89412-810 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V China/Aust 10" 302510040 89412-818 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V India 8" 302580070 89412-874 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet 230V India 10" 302510070 89412-882 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V India 8" 302581170 89412-880 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V Schuko 10" 302510130 89412-758 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V India 8" 302580170 89412-878 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V UK 8" 302580150 89412-686 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V UK 10" 302510150 89412-694 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V China/Aust 8" 302580140 89412-814 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V China/Aust 10" 302510140 89412-822 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 100V Japan 8" 302580120 89412-638 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V Schuko 8" 302580130 89412-750 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand 230V India 10" 302510170 89412-886 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V UK 8" 302581150 89412-688 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V Schuko 8" 302581130 89412-752 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V China/Aust 8" 302581140 89412-816 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V China/Aust 10" 302511140 89412-824 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V UK 10" 302511150 89412-696 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V Schuko 10" 302511130 89412-760 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 230V India 10" 302511170 89412-888 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with Telescoping Base Stand, UV Light, Service Fixtures, and Pass Through 100V Japan 8" 302581120 89412-640 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V Schuko 8" 302581030 89412-748 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V China/Aust 8" 302581040 89412-812 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 100V Japan 8" 302581020 89412-636 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V UK 8" 302581050 89412-684 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V UK 10" 302511050 89412-692 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V China/Aust 10" 302511040 89412-820 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V India 8" 302581070 89412-876 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V India 10" 302511070 89412-884 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
5' Purifier Logic+ A2 Biosafety Cabinet with UV Light, Service Fixtures, and Pass Through 230V Schuko 10" 302511030 89412-756 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product is energy and/or water efficient
ACCESSORIES
Expand All / Collapse All
Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Replacement Exhaust HEPA Filter for 5' Purifier Logic 3838502 97006-996 Each Retrieving
Replacement Supply HEPA Filter for 5' Purifier Logic 3838402 97007-024 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SERVICES
Services: VWRCATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST™@vwr.com for more details. You can also visit https://vwr.com/equip_services.
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty