UV Transilluminator Bulb, 365 nm, for myECL™ Imager, Thermo Scientific
Supplier: Thermo Scientific
62255 62258
PI62258EA 71.4 USD
PI62258 PI62255
UV Transilluminator Bulb, 365 nm, for myECL™ Imager, Thermo Scientific
Photodocumentation Systems
This lamp bulb with ultraviolet emission (365nm) is used to replace the pre-installed 306nm UV bulbs in the Thermo Scientific myECL Imager with long-wave 365nm bulbs; all six lamps must be the same bulb type (365nm or 306nm).
Ordering information: For Research Use Only. Not for use in diagnostic procedures.