Exotoxin A, List Biological
160
102946-476EA 320 USD
102946-476
Exotoxin A, List Biological
Exotoxin A is one of the toxic proteins released by pathogenic strains of Pseudomonas aeruginosa
It is secreted as a proenzyme with a molecular weight of 66,000 daltons. Exotoxin A is translocated into susceptible mammalian cells.