Sintered Glass Fiber Filter Discs, Porosity E, Ace Glass Incorporated
Supplier: Ace Glass
7176-136 7176-166 7176-36 7176-46 7176-146 7176-61 7176-156 7176-10 7176-126 7176-71 7176-05 7176-26 7176-16 7176-41 7176-105 7176-116 7176-56 7176-21 7176-31 7176-51 7176-81
89057-790EA 29.76 USD
89057-790 89057-964 89057-886 89057-974 89057-984 89057-852 89057-994 89057-814 89057-748 89057-802 89057-934 89057-758 89057-944 89057-768 89057-900 89057-954 89057-778 89057-828 89057-838 89057-826 89058-004
Sintered Glass Fiber Filter Discs, Porosity E, Ace Glass Incorporated
Filters Glass Fiber Filters
These glass fiber filter discs last longer and are more abrasion resistant.
  • Superior in performance to granular type discs
  • Resistant to thermal shock and chemical attack
  • Fabricated from borosilicate glass
Available in a complete range of porosities and diameters, these discs can be used in the manufacture of a wide variety of special apparatus.