N-Acetyl-5-hydroxytryptamine ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms: N-(2-(5-Hydroxy-1H-indol-3-yl)ethyl)acetamide
550-208-M500
89152-346EA 145.59 USD
89152-346
N-Acetyl-5-hydroxytryptamine ≥98% (by HPLC)
N-Acetyl-5-hydroxytryptamine
Hydroxyindole-O-methyltransferase inhibitor
Metabolite of serotonin (Prod. No. ALX-550-328). Precursor of melatonin (Prod. No. BML-NS520). Inhibitor of hydroxyindole-O-methyltransferase.