Uranyl acetate 1% in aqueous solution Ready-to-Use

Supplier: Electron Microscopy Sciences
Warning

22400-1
102092-282EA 77.76 USD
102092-282
Uranyl acetate 1% in aqueous solution Ready-to-Use
Uranyl acetate

Pre-made and ready to use uranyl acetate solution are avaialable with concentrations of 1%, 2%, 3%, or 4%.


Other concentrations are available upon request.

Formula: C₄H₈O₆U
Boiling Pt: 100 °C (212 °F)
Storage Temperature: Ambient
CAS Number: 541-09-3
UN: 2910
ADR: 7,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR