Oil red O
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
Sudan Red 5B
, CI 26125
, Solvent Red 27
19056
102091-000EA
32.08
USD
102091-000
Oil red O
Oil Red O
Oil red O is lysochrome (fat soluble dye) used demonstrate fat or lipids in fresh tissue sections.
The polyazo group of dyes include the oil red, sudan red and the sudan blacks dyes are interchangeable.
Formula:
C₂₆H₂₄N₄O MW: 408.51 g/mol |
MDL Number:
MFCD00003898 CAS Number: 1320-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...