Leishman's eosin-methylene blue

Supplier: Electron Microscopy Sciences

Synonyms: Eosin and methylene blue LeishmanEosin-Methylene blue dye LeishmanLeishman stainLeishman's stain (eosin methylene blue compound)Leishman's stain

26086-05
101410-986EA 108.42 USD
101410-986
Leishman's eosin-methylene blue
Leishman's Eosin-Methylene blue

Lieshman’s Stain and Working Buffer

MDL Number: MFCD00131498
CAS Number: 12627-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR