Thionin (Chloride) 0.5% in 20% Alcohol

Supplier: Electron Microscopy Sciences

Synonyms: 3,7-Diaminophenazathionium chlorideLauth's VioletThioninKatalysin

21910
101410-828EA 97.62 USD
101410-828
Thionin (Chloride) 0.5% in 20% Alcohol
Thionin (Chloride)
Formula: C₁₂H₁₀ClN₃S
MW: 263.74 g/mol
MDL Number: MFCD00078211
CAS Number: 581-64-6
https://us.vwr.com/stibo/web/std.lang.all/49/17/28984917.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR