Tannic acid 10% in aqueous solution

Supplier: Electron Microscopy Sciences

Synonyms: TanninGallotannin

21702-10
101410-818EA 58.23 USD
101410-818
Tannic acid 10% in aqueous solution
Tannic acid

Tannic Acid solution Electron Microscopy Sciences is available in concentrations of 5.0% aqueous and 10.0% aqueous.

Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
MDL Number: MFCD00066397
CAS Number: 1401-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR