Biebrich scarlet 0.2% in 1% acetic acid

Supplier: Electron Microscopy Sciences

Synonyms: Bonceau BSAcid Red 66Double Scarlet BSFCroceine ScarletCI 26905Biebrich Scarlet sodium salt

26033-02
101410-896EA 48.82 USD
101410-896
Biebrich scarlet 0.2% in 1% acetic acid
Biebrich Scarlet

Biebrich Scarlet solution Electron Microscopy Sciences is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology. Available in concentrations of 0.2% or 1.0% in 1% acetic acid, 1% aqueous, or with acid fushin.

Formula: C₂₂H₁₄N₄Na₂O₇S₂
MW: 556.49 g/mol
MDL Number: MFCD00003891
CAS Number: 4196-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR