Giemsa's stain stock solution acc. to Price stain for hematology

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Giemsa's azur-eosin-methylene blueAzure mixture sicc. Giemsa stainAzur-Eosin-Methylene blue acc. Giemsa

26156-01
100504-552EA 50.95 USD
100504-552
Giemsa's stain stock solution acc. to Price stain for hematology
Giemsa's stain

Price’s Giemsa Stain is used for hematologic elements. Stain results - Malarial Parasites: Blue, Tissue Nuclei: Blue, Bacteria: Dark Blue, Malarial Pigment: Black, Schistosonic egg shells: Blue, Collagen, etc.: Pale Pink, Erthrocytes: Pink Rose..


The Price's method includes the use of the following reagents:


Phosphate buffer pH 7.0 100504-552
0.2% Acetic acid 100504-554
2% alcoholic iodine  100504-992


References: AFIP Manual of Histological Staining Methods, 3rd ed., L. Luna: New York: McGraw Hill Publications, c. 1968, p. 127. Price, D.L., Mil. Med., 133: 363-367, 1968.

Flash Pt: 11 °C
MDL Number: MFCD00081642
CAS Number: 51811-82-6
UN: 1230
ADR: 3 (6.1),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR