Wright's stain, certified stain for hematology (CI DcWr-38)

Supplier: Electron Microscopy Sciences

Synonyms: Eosin Methylene blue solution according to WrightWright's eosin methylene blue

22720
100504-340EA 58.33 USD
100504-340
Wright's stain, certified stain for hematology (CI DcWr-38)
Wright's stain

For differentiation of blood corpuscles


For differentiation of blood corpuscles. Used with brilliant cresyl blue for staining platelets and reticulum of immature red cells. For staining blood and bone marrow films.

MDL Number: MFCD00082143
CAS Number: 68988-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR