Sodium ethoxide

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Sodium ethylateSodium ethanolate

21150
100504-280EA 13.07 USD InStock
100504-280
Sodium ethoxide
Sodium ethoxide

Used as an agent for flattening thick sections (0.5 to 2.0 micron) of epoxy embedded material (Felix Sierra, Christa Hegele-Hartung and Henning M. Beier (1990)).


An improved technique for flattening semithin sections of epoxy-embedded tissues used for high resolution light microscopy (J. of Microscopy Vol 159, Pt 1, (July 1990), pp. 95-98).

Formula: C₂H₅NaO
MW: 68,05 g/mol
Melting Pt: 260 °C
Density: 0,88 g/cm³ (7.344 lbs/gal.)
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR