Pyronin G ≥99%, certified (C.I. 45005 - C.N. DcPy-2)

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Pyronine Y3,6-Bis(dimethylamino)xanthylium chloridePyronin Y

19560
100504-198EA 116.82 USD
100504-198
Pyronin G ≥99%, certified (C.I. 45005 - C.N. DcPy-2)
Pyronine G

Cudder's method combined gram/pappenheim stain for gonorrheal pus. Kurnick's method used with methyl green which stains liver cells. For staining protein in the diazosulfanilic acid technique; a substitute for azur A. can be used for pyronin B.


Solubility : 9.0% Water; 0.5% Alcohol; 1.85% Cellosolve; 4.35% Glycol; 0.0% Xylene

Formula: C₁₇H₁₉ClN₂O
MW: 302.8 g/mol
Melting Pt: 250…260 °C (482…500 °F)
Density: 0.64 g/cm³ (5.341 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0
https://us.vwr.com/stibo/web/std.lang.all/48/89/28984889.jpg

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR