Polyvinyl alcohol, M.W. 15000

Supplier: Electron Microscopy Sciences

Synonyms: Mowiol®PVOHPVA

19800
100504-222EA 32.18 USD
100504-222
Polyvinyl alcohol, M.W. 15000
Polyvinyl alcohol

A water-miscible polymer used as an embedding medium for electron microscopy.

Formula: [-CH₂CH(OH)-]n
Density: 1.269 g/cm³ (10.59 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5
https://us.vwr.com/stibo/web/std.lang.all/48/80/28984880.jpg

Order Now

Specification Test Results

Degree of hydrolyzation 86-89 mol%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR