Periodic acid ≥99%
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Orthoperiodic acid
, ortho-Periodic acid
19325
100504-182EA
53.53
USD
100504-182
Periodic acid ≥99%
Periodic acid
Ainsworth, S.K. et al. (1972). Alkaline bismuth reagent for high resolution ultrastructural demonstration of periodic-reactive sites (J. Histochem. Cytochem., 20:995).
Mowry, R.W. (1958)). Improved procedure for the staining of acid polysaccharides by Mulle.
Formula:
H₅IO₆ MW: 227.94 g/mol Melting Pt: 122 °C (252 °F) (Zers.) Density: 1.4 g/cm³ (11.683 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00011344 CAS Number: 10450-60-9 UN: 3085 ADR: 5.1,II |
Specification Test Results
Assay |
|
Heavy Metals | 0.005% |
Insoluble Matters | 0.01% |
Iron | 0.003% |
Other Halogen (Cl) | 0.01% |
Residue after ignition | 0.01% |
Sulfate | 0.01% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...