Parlodion (Pyroxylin Purified Strips)

Supplier: Electron Microscopy Sciences
Warning

19220
100504-176EA 342.61 USD
100504-176
Parlodion (Pyroxylin Purified Strips)
Parlodion (Pyroxylin Purified Strips)

Highly purified from cellulose nitrate; specially prepared for embedding tissue for sectioning and for preparing semi-permeable membranes.

Formula: C₁₂H₁₆N₄O₁₈
MW: 504.28 g/mol
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
CAS Number: 9004-70-0
UN: 2059
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR