Butyl methacrylate monomer stabilized

Supplier: Electron Microscopy Sciences
Warning

Synonyms: 2-Methylacrylic acid, butyl esterMethacrylic acid butyl estern-Butyl Methacrylate, MonomerButyl 2-methacrylate

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 10 ppm

12100
100505-018EA 25.42 USD
100505-018
Butyl methacrylate monomer stabilized
Butyl methacrylate
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Melting Pt: –75 °C (–103 °F)
Density: 0.9836 g/cm³ (8.208 lbs/gal.)
Flash Pt: 54 °C (129 °F)
MDL Number: MFCD00009444
CAS Number: 97-88-1
UN: 2227
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/48/59/28984859.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR