N,N-Dimethylaniline

Supplier: Electron Microscopy Sciences
Danger

13360
100503-830EA 13.9 USD
100503-830
N,N-Dimethylaniline
N,N-Dimethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 194 °C (381 °F)
Melting Pt: 2.45 °C (36 °F)
Density: 0.96 g/cm³ (8.011 lbs/gal.)
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008304
CAS Number: 121-69-7
UN: 2253
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/48/58/28984858.jpg

Order Now

Specification Test Results

Specific Gravity 0.96 g/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR