γ-(N-Morpholino)propanesulfonic acid (MOPS) biological buffer pH 6.5 - 7.9

Supplier: Electron Microscopy Sciences
Warning

Synonyms: 3-(N-Morpholino)propanesulphonic acid3-Morpholinopropane-1-sulfonic acid3-Morpholinopropane-1-sulphonic acid3-(N-Morpholino)propanesulfonic acid

18900
100504-114EA 116.51 USD
100504-114
γ-(N-Morpholino)propanesulfonic acid (MOPS) biological buffer pH 6.5 - 7.9
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: 277…280 Zers °C (531…536 Zers °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006183
CAS Number: 1132-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR