Lucifer yellow CH dilithium salt

Supplier: Electron Microscopy Sciences

Synonyms: 6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulphonic acid dilithium saltLY CH lithium salt6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulfonic acid dilithium salt

17950
100504-948EA 631.48 USD
100504-948
Lucifer yellow CH dilithium salt
Lucifer yellow CH dilithium salt

A technique producing an electron dense osmiophilic polymer when Lucifer yellow is injected into cells. Lucifer yellow also can be viewed in living cells as a dye marker and is more sensitive than any other dye.


Cell Tissue Res., 204, 37 (1979) J. Histochem Cystochem, 30, 189 (1982) Science, 217, 953 (1982); 215, 1641 (1982); 214, 801 (1981); 225, 851 (1984); Cell, 14, 741 (1978).
A fluorescent dye. Nature 292, 17 (1981).

Formula: C₁₃H₉N₅O₉S₂Li₂
MW: 457.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006922
CAS Number: 67769-47-5
https://us.vwr.com/stibo/web/std.lang.all/48/44/28984844.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR