Lanthanum(III) nitrate hexahydrate, Reagent Grade

Supplier: Electron Microscopy Sciences
Warning

17300
100504-066EA 188.86 USD
100504-066
Lanthanum(III) nitrate hexahydrate, Reagent Grade
Lanthanum (III) nitrate hexahydrate

A metal stain, intracellular tracer (Lane, N,J. and Treherne, J.E. (1970). Lanthanum staining of neurotubules in axons from cockroach ganglia. J. Cell Sci., 7:217), (Lane, N.J. (1972) Fine structure of a lipidopteran nervous system and its accessibility to peroxidase and lanthanum. Z. Zellforsch., 131:205).

Formula: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay Min. 99.9%
Calcium 0.001%
Magnesium 0.0001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR