(±)-2-Methyl-2,4-pentanediol 99% ACS
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
(±)-2,4-Dihydroxy-2-methyl pentane
, 2-Methylpentane-2,4-diol
, Hexylene Glycol
, Hexylene glycol
16750
16740
102090-984CS
170.06
USD
102090-984
100504-036
(±)-2-Methyl-2,4-pentanediol 99% ACS
(±)-2-Methyl-2,4-pentanediol
Mixed with water in all proportions. Used as a dehydrating agent for fat tissues.
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Boiling Pt: 198.5 °C (389 °F) Melting Pt: –50 °C (–58 °F) Density: 0.922 g/cm³ (7.694 lbs/gal.) Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004547 CAS Number: 107-41-5 |
Specification Test Results
Assay | 99% |
IR Spectrum | To Pass Test |
Refractive Index @20°C | 0.921-0.923 |
Specific gravity @25°C | 0.923 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...