Epon-815 resin
Supplier: Electron Microscopy Sciences
14910
100503-880EA
60.32
USD
100503-880
Epon-815 resin
Epon-815 resin
CAS NA
Specific Gravity: 1.13
An aromatic resin.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...