Divinylbenzene (mixture of isomers)
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
Divinylbenzene
13450
100504-936EA
21.46
USD
100504-936
Divinylbenzene (mixture of isomers)
Divinylbenzene (mixture of isomers)
A cross-linking monomer. Inhibited with up to 1500 ppm p-tert-butyl catechol.
Formula:
C₁₀H₁₀ MW: 130.19 g/mol Boiling Pt: 200 °C (392 °F) Melting Pt: –67 °C (–89 °F) Density: 0.915 g/cm³ (7.636 lbs/gal.) Flash Pt: 64 °C (147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01778656 CAS Number: 1321-74-0 |
Specification Test Results
Specific Gravity | 0.91 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...