CytoPrep™ Fixative, Electron Microscopy Science
64145-12 64145-01
100501-706EA 50.36 USD
100501-706 100500-436
CytoPrep™ Fixative, Electron Microscopy Science
CytoPrep fixative forms a clear protective coating over specimens while fixing cells. Ideal for cytological smears and it dries within 4 to 6 minutes.
  • Good for safe storage and transportation of slides
  • Water soluble and contains no chlorofluorocarbons
CytoPrep™ Fixative forms a clear protective coating over specimens while fixing cells. It is ideal for cytological smears and it dries within 4 to 6 minutes. Good for safe storage and transportation of slides. It is water soluble and contains no chlorofluorocarbons. Comes in a 4 oz. spray can.
Packaging: Packed in a 4 oz. spray can.