Benzoin UV Catalyst

Supplier: Electron Microscopy Sciences

Synonyms: 2-Hydroxy-2-phenylacetophenone

11270
100503-682EA 20.61 USD
100503-682
Benzoin UV Catalyst
Benzoin
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 344 °C (651 °F)
Density: 0.5 g/cm³ (4.173 lbs/gal.)
MDL Number: MFCD00004496
CAS Number: 119-53-9
https://us.vwr.com/stibo/web/std.lang.all/47/84/28984784.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR