Benzoin methyl ether ≥98% UV Catalyst

Supplier: Electron Microscopy Sciences
Danger

Synonyms: 2-Methoxy-1,2-diphenylethanone1,2-Diphenyl-2-methoxyethan-1-one

11290
100503-686EA 72.18 USD
100503-686
Benzoin methyl ether ≥98% UV Catalyst
Benzoin methyl ether
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00008492
CAS Number: 3524-62-7
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/47/86/28984786.jpg

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR